α,α,α,3-Tetrafluorotoluene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15055-14
A15055-22
CAAAA15055-14EA
67.62
CAD
CAAAA15055-14
CAAAA15055-22
α,α,α,3-Tetrafluorotoluene 99%
α,α,α,3-Tetrafluorotoluene
Purity: 99%
CAS No.: 401-80-9
Molecular Formula: C7H4F4
Formula Weight: 164.10
Flash Point: 7°(44°F)
Refractive Index: 1.4010
Boiling Point: 101-102°
Beilstein Reference: 2045664
UN No.: 1993
MDL No.: MFCD00000382
Formula:
C₇H₄F₄ Boiling Pt: 101…102 °C Melting Pt: –81.5 °C Density: 1.303 Flash Pt: 7 °C (44 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000382 CAS Number: 401-80-9 EINECS: 206-933-1 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...