Piperazine, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Danger

A15049-22 A15049-36
CAAAAA15049-A1EA 177.72 CAD
CAAAAA15049-A1 CAAAA15049-22 CAAAA15049-36 CAAAAA15049-61
Piperazine, anhydrous 99%
Piperazine
Formula: C₄H₁₀N₂
Boiling Pt: 146…148 °C
Melting Pt: 108…113 °C
Density: 1.110
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005953
CAS Number: 110-85-0
EINECS: 203-808-3
UN: 2579
ADR: 8,III
Merck Index: 14,7464

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR