Ethyl pyruvate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-oxopropanoate, 2-Oxopropionic acid ethyl ester, α-Ketopropionic acid ethyl ester, Pyruvic acid ethyl ester, 2-Ketopropionic acid ethyl ester
A15045-36
A15045-18
A15045-30
CAAAAA15045-36EA
429.76
CAD
CAAAAA15045-36
CAAAA15045-18
CAAAA15045-30
Ethyl pyruvate 98%
Ethyl pyruvate
Purity: 98%
CAS No.: 617-35-6
Molecular Formula: C5H8O3
Formula Weight: 116.12
Storage Temperature: 2°C to 8°C
Flash Point: 45°(113°F)
Refractive Index: 1.4050
Odor: Pleasant
Melting Point: -58°
Boiling Point: 143-145°
Beilstein Reference: 1071466
Merck Reference: 14,8021
UN No.: 3272
MDL No.: MFCD00009123
Formula:
C₅H₈O₃ MW: 116.12 g/mol Boiling Pt: 143…145 °C Melting Pt: -50 Density: 1.050 Flash Pt: 45 °C (113 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009123 CAS Number: 617-35-6 EINECS: 210-511-2 UN: 3272 ADR: 3 III Merck Index: 14,8021 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...