Sodium octadecyl sulfate, (max. 7% H₂O) 98% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium Stearyl sulfate, n-Octadecyl sulfate, sodium salt, Sodium Stearyl sulphate, Octadecyl sulfuric acid salt, Sodium n-octadecylsulfate, n-Octadecyl sulphate, sodium salt, Sodium n-octadecylsulphate, Octadecyl sulphuric acid salt

A15042-03 A15042-06 A15042-14
CAAAA15042-03EA 114.23 CAD
CAAAA15042-03 CAAAA15042-06 CAAAA15042-14
Sodium octadecyl sulfate, (max. 7% H₂O) 98% (dry weight)
Sodium octadecyl sulphate

Purity: 98% CAS No.: 1120-04-3 Molecular Formula: C18H37NaO4S Formula Weight: 372.55 Melting Point: 176-180° Beilstein Reference: 3640871

Formula: C₁₈H₃₇NaO₄S
MW: 372.55 g/mol
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00007469
CAS Number: 1120-04-3
EINECS: 214-295-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR