3,4,5-Trimethoxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

A15038-36 A15038-14 A15038-22
CAAAAA15038-36EA 485.06 CAD
CAAAAA15038-36 CAAAA15038-14 CAAAA15038-22
3,4,5-Trimethoxybenzaldehyde 98%
3,4,5-Trimethoxybenzaldehyde

Purity: 98% CAS No.: 86-81-7 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 73-77° Boiling Point: 163-165°/10mm MDL No.: MFCD00003364

Formula: C₁₀H₁₂O₄
Boiling Pt: 163…165 °C/10 mm
Melting Pt: 73…77 °C
Density: 1.367 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003364
CAS Number: 86-81-7
EINECS: 201-701-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR