Salicylaldehyde ethyl ether 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15035-22
A15035-18
CAAAAA15035-22EA
197.5
CAD
CAAAAA15035-22
CAAAA15035-18
CAAAA15035-30
Salicylaldehyde ethyl ether 97+%
Salicylaldehyde ethyl ether
Purity: 97+%
CAS No.: 613-69-4
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 91°(195°F)
Refractive Index: 1.5430
Boiling Point: 249°
Beilstein Reference: 1937270
MDL No.: MFCD00003316
Formula:
C₉H₁₀O₂ Boiling Pt: 249 °C Melting Pt: 20…22 °C Density: 1,077 Flash Pt: 91 °C(195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003316 CAS Number: 613-69-4 EINECS: 210-349-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...