L(+)-Glutamic acid 99+%
Supplier: Thermo Scientific Chemicals
A15031-36
A15031-0B
A15031-0I
A15031-30
CAAAAA15031-36EA
87.67
CAD
CAAAAA15031-36
CAAAA15031-0B
CAAAA15031-0I
CAAAA15031-30
L(+)-Glutamic acid 99+%
L(+)-Glutamic acid
Purity: 99+%
CAS No.: 56-86-0
Molecular Formula: C5H9NO4
Formula Weight: 147.13
Physical Form: Cryst powder
Density: 1.525
Appearance: White
Melting Point: 189-192° dec.
Beilstein Reference: 1723801
Merck Reference: 14,4469
Optical Rotation: +31° (c=5 in 5N HCl)
MDL No.: MFCD00002634
Formula:
HO₂CCH₂CH₂CH(NH₂)CO₂H Boiling Pt: 325 °C (1013 hPa) Melting Pt: 189…192 °C (dec.) Density: 1.525 Storage Temperature: Ambient |
MDL Number:
MFCD00002634 CAS Number: 56-86-0 EINECS: 200-293-7 Merck Index: 14,4469 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...