(4-Hydroxyphenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A15018-14 A15018-22 A15018-36
CAAAA15018-14EA 74.73 CAD
CAAAA15018-14 CAAAA15018-22 CAAAA15018-36
(4-Hydroxyphenyl)acetic acid 99%
(4-Hydroxyphenyl)acetic acid

Purity: 99% CAS No.: 156-38-7 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 148-150° Beilstein Reference: 1448766

Formula: (C₆H₄(OH))CH₂COOH
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004347
CAS Number: 156-38-7
EINECS: 205-851-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR