2,6-Dimethylbenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15014-03
A15014-06
A15014-14
CAAAA15014-03EA
57.2
CAD
CAAAA15014-03
CAAAA15014-06
CAAAA15014-14
2,6-Dimethylbenzoic acid 98+%
2,6-Dimethylbenzoic acid
Purity: 98+%
CAS No.: 632-46-2
Molecular Formula: C9H10O2
Formula Weight: 150.18
Odor: Odorless
Melting Point: 114-117°
Beilstein Reference: 1863791
MDL No.: MFCD00002483
Formula:
C₉H₁₀O₂ Boiling Pt: 155 - 160 °C (17 -rr) Melting Pt: 114…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002483 CAS Number: 632-46-2 EINECS: 211-177-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...