Ethyl-4-ethoxybenzoate 98%
Supplier: Thermo Scientific Chemicals
A15010-14
A15010-22
CAAAA15010-14EA
142.83
CAD
CAAAA15010-14
CAAAA15010-22
Ethyl-4-ethoxybenzoate 98%
Ethyl 4-ethoxybenzoate
Purity: 98%
CAS No.: 23676-09-7
Molecular Formula: C11H14O3
Formula Weight: 194.23
Flash Point: 137°(278°F)
Refractive Index: 1.5180
Melting Point: 9°
Boiling Point: 145-150°/13mm
Beilstein Reference: 2692236
MDL No.: MFCD00009116
Formula:
C₁₁H₁₄O₃ Boiling Pt: 145…150 °C/13 mm Melting Pt: 9 °C Density: 1.071 Flash Pt: 137 °C(278 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009116 CAS Number: 23676-09-7 EINECS: 245-818-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...