Benzyl isothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzylisothiocyanate

A15008-06 A15008-14 A15008-22
CAAAA15008-06EA 52.93 CAD
CAAAA15008-06 CAAAA15008-14 CAAAA15008-22
Benzyl isothiocyanate 98%
Benzyl isothiocyanate

Purity: 98% CAS No.: 622-78-6 Molecular Formula: C6H5CH2NCS Formula Weight: 149.22 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.6030 Odor: Pungent Boiling Point: 242-243° Beilstein Reference: 386135 UN No.: 2922 MDL No.: MFCD00004819

Formula: C₈H₇NS
MW: 149.22 g/mol
Boiling Pt: 243
Melting Pt: 24
Density: 1.125
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004819
CAS Number: 622-78-6
EINECS: 210-753-9
UN: 2206

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR