Ethyl levulinate 98%
Supplier: Thermo Scientific Chemicals
Warning
|
A15001-22
A15001-36
A15001-18
CAAAAA15001-22EA
80.58
CAD
CAAAAA15001-22
CAAAAA15001-36
CAAAA15001-18
Ethyl levulinate 98%
Ethyl levulinate
Purity: 98%
CAS No.: 539-88-8
Molecular Formula: C7H12O3
Formula Weight: 144.17
Flash Point: 94°(201°F)
Refractive Index: 1.4220
Boiling Point: 203-205°
Beilstein Reference: 507641
Merck Reference: 14,3819
MDL No.: MFCD00009209
Formula:
C₇H₁₂O₃ Boiling Pt: 203…205 °C Density: 1.014 Flash Pt: 94 °C(201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009209 CAS Number: 539-88-8 EINECS: 208-728-2 Merck Index: 14,3819 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...