4-Methoxybenzylamine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-(Aminomethyl)anisole, p-Anisylamine
A14999-36
A14999-14
A14999-22
CAAAAA14999-36EA
1281.38
CAD
CAAAAA14999-36
CAAAA14999-14
CAAAA14999-22
4-Methoxybenzylamine 98+%
4-Methoxybenzylamine
Purity: 98+%
CAS No.: 2393-23-9
Molecular Formula: C8H11NO
Formula Weight: 137.18
Flash Point: 114°(237°F)
Refractive Index: 1.5460
Odor: Amine-like
Melting Point: -10°
Boiling Point: 236-237°
Beilstein Reference: 508206
UN No.: 2735
MDL No.: MFCD00008122
Formula:
C₈H₁₁NO MW: 137.18 g/mol Boiling Pt: 236…237 °C Melting Pt: –10 °C Density: 1.053 Flash Pt: 114 °C (237 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008122 CAS Number: 2393-23-9 EINECS: 219-247-2 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...