Bromocyclopropane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14996-30
A14996-09
A14996-18
CAAAAA14996-30EA
1896
CAD
CAAAAA14996-30
CAAAA14996-09
CAAAA14996-18
Bromocyclopropane 99%
Purity: 99%
CAS No.: 4333-56-6
Molecular Formula: C3H5Br
Formula Weight: 120.98
Storage Temperature: 2°C to 8°C
Flash Point: -6°(21°F)
Refractive Index: 1.4600
Odor: Recognizable
Boiling Point: 68-70°
Beilstein Reference: 1900287
UN No.: 1993
MDL No.: MFCD00001271
Formula:
C₃H₅Br Boiling Pt: 68…70 °C Density: 1.515 Flash Pt: –6 °C (21 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001271 CAS Number: 4333-56-6 EINECS: 224-375-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...