1-Dodecene 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14992-22
A14992-36
CAAAA14992-22EA
115.02
CAD
CAAAA14992-22
CAAAA14992-36
1-Dodecene 96%
1-Dodecene
Purity: 96%
CAS No.: 112-41-4
Molecular Formula: C12H24
Formula Weight: 168.32
Flash Point: 73°(163°F)
Refractive Index: 1.4300
Melting Point: -37°
Boiling Point: 212-213°
Beilstein Reference: 1699848
MDL No.: MFCD00008961
Formula:
C₁₂H₂₄ Boiling Pt: 212…213 °C Melting Pt: –37 °C Density: 0.758 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008961 CAS Number: 112-41-4 EINECS: 203-968-4 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...