4-Amino-3,5-dichlorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A14982-06 A14982-14
CAAAA14982-06EA 57.04 CAD
CAAAA14982-06 CAAAA14982-14
4-Amino-3,5-dichlorobenzoic acid 98%
4-Amino-3,5-dichlorobenzoic acid

Purity: 98% CAS No.: 56961-25-2 Molecular Formula: C7H5Cl2NO2 Formula Weight: 206.03 Melting Point: ca 290° dec. Beilstein Reference: 2805751

Formula: C₇H₅Cl₂NO₂
MW: 206.03
Melting Pt: ∼ 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00017091
CAS Number: 56961-25-2
EINECS: 260-468-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR