2-Fluorophenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14979-14
A14979-22
CAAAA14979-14EA
95.43
CAD
CAAAA14979-14
CAAAA14979-22
CAAAA14979-36
2-Fluorophenol 98%
2-Fluorophenol
Purity: 98%
CAS No.: 367-12-4
Molecular Formula: C6H5FO
Formula Weight: 112.10
Flash Point: 46°(114°F)
Refractive Index: 1.5140
Odor: Phenol-like
Melting Point: 14-16°
Boiling Point: 150-152°
Beilstein Reference: 1905112
UN No.: 2924
MDL No.: MFCD00002155
Formula:
C₆H₅FO Boiling Pt: 150…152 °C Melting Pt: 14…16 °C Density: 1.215 Flash Pt: 46 °C (114 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002155 CAS Number: 367-12-4 EINECS: 206-681-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...