(4-Chlorophenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Chlorophenyl)acetic acid, p-Chlorophenylacetic Acid

A14974-0B A14974-18 A14974-30
CAAAA14974-0BEA 1173.94 CAD
CAAAA14974-0B CAAAA14974-18 CAAAA14974-30
(4-Chlorophenyl)acetic acid 98%
(4-Chlorophenyl)acetic acid

Purity: 98% CAS No.: 1878-66-6 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Odor: Odorless Melting Point: 103-107° Beilstein Reference: 1072816 MDL No.: MFCD00004344

Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Boiling Pt: 292
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00004344
CAS Number: 1878-66-6
EINECS: 217-521-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR