1,2,3,4-Tetrahydronaphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning

A14962-0B A14962-0I A14962-30
CAAAA14962-0BEA 98.28 CAD
CAAAA14962-0B CAAAA14962-0I CAAAA14962-30
1,2,3,4-Tetrahydronaphthalene 97%
1,2,3,4-Tetrahydronaphthalene

Purity: 97% CAS No.: 119-64-2 Molecular Formula: C10H12 Formula Weight: 132.21 Flash Point: 77°(171°F) Refractive Index: 1.5400 Melting Point: -36° Boiling Point: 206-207° Beilstein Reference: 1446407 Merck Reference: 14,9221 UN No.: 3082 MDL No.: MFCD00001733

Formula: C₁₀H₁₂
MW: 132.21 g/mol
Boiling Pt: 206…207 °C
Melting Pt: –36 °C
Density: 0.966
Flash Pt: 77 °C (171 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001733
CAS Number: 119-64-2
EINECS: 204-340-2
UN: 3082
ADR: 9,III
Merck Index: 14,9221

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR