Bicine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine
A14957-0B
A14957-18
A14957-30
CAAAA14957-0BEA
711
CAD
CAAAA14957-0B
CAAAA14957-18
CAAAA14957-30
Bicine 99%
Bicine
Good's buffers
Purity: 99%
CAS No.: 150-25-4
Molecular Formula: C6H13NO4
Formula Weight: 163.17
pH: 4-5
Melting Point: ca 191° dec.
Beilstein Reference: 1769362
Merck Reference: 14,1201
MDL No.: MFCD00004295
Good's buffers
Formula:
C₆H₁₃NO₄ MW: 163.17 g/mol Melting Pt: ca 191 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004295 CAS Number: 150-25-4 EINECS: 205-755-1 Merck Index: 14,1201 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...