4-Bromobutyl phenyl ether 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14956-06
A14956-14
A14956-22
CAAAA14956-06EA
84.53
CAD
CAAAA14956-06
CAAAA14956-14
CAAAA14956-22
4-Bromobutyl phenyl ether 97%
4-Bromobutyl phenyl ether
Purity: 97%
CAS No.: 1200-03-9
Molecular Formula: C6H5O(CH2)4Br
Formula Weight: 229.12
Flash Point:
Formula:
(C₆H₅)O(CH₂)₄Br Boiling Pt: 153…154 °C/18 mm Melting Pt: 40…43 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000262 CAS Number: 1200-03-9 EINECS: 214-849-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...