4-Chloro-3-nitrocinnamic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4-Chloro-3-nitrophenyl)prop-2-enoic acid, 3-(4-Chloro-3-nitrophenyl)acrylic acid

A14954-06 A14954-22
CAAAA14954-06EA 39.97 CAD
CAAAA14954-06 CAAAA14954-14 CAAAA14954-22
4-Chloro-3-nitrocinnamic acid 98%
4-Chloro-3-nitrocinnamic acid

Purity: 98% CAS No.: 20797-48-2 Molecular Formula: C9H6ClNO4 Formula Weight: 227.61 Melting Point: 186-188° MDL No.: MFCD00063311

Formula: C₉H₆ClNO₄
Melting Pt: 186…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00063311
CAS Number: 20797-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR