α,α,α-Trifluoro-o-toluonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14951-06
A14951-09
CAAAAA14951-06EA
68.57
CAD
CAAAAA14951-06
CAAAA14951-09
CAAAA14951-18
α,α,α-Trifluoro-o-toluonitrile 97%
α,α,α-Trifluoro-o-toluonitrile
Purity: 97%
CAS No.: 447-60-9
Molecular Formula: C8H4F3N
Formula Weight: 171.12
Flash Point: 90°(194°F)
Refractive Index: 1.4632
Melting Point: 17-18°
Boiling Point: 204-206°
Beilstein Reference: 2048152
UN No.: 3276
MDL No.: MFCD00001791
Formula:
C₈H₄F₃N Boiling Pt: 204…206 °C Melting Pt: 17…18 °C Density: 1.294 Flash Pt: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001791 CAS Number: 447-60-9 EINECS: 207-184-3 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...