4-Benzyloxybenzyl alcohol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14949-09
A14949-18
CAAAA14949-09EA
70.94
CAD
CAAAA14949-09
CAAAA14949-18
CAAAA14949-30
4-Benzyloxybenzyl alcohol 98+%
4-Benzyloxybenzyl alcohol
Purity: 98+%
CAS No.: 836-43-1
Molecular Formula: C14H14O2
Formula Weight: 214.26
Melting Point: 84-88°
Beilstein Reference: 1877819
Formula:
C₁₄H₁₄O₂ Melting Pt: 84…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004654 CAS Number: 836-43-1 EINECS: 212-649-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...