Methyl propionate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14944-0F
A14944-AE
A14944-AP
CAAAA14944-0FEA
647.8
CAD
CAAAA14944-0F
CAAAA14944-AE
CAAAA14944-AP
Methyl propionate 99%
Methyl propionate
Purity: 99%
CAS No.: 554-12-1
Molecular Formula: CH3CH2CO2CH3
Formula Weight: 88.11
Flash Point: -2°(28°F)
Physical Form: Liquid
Density: 0.915
Refractive Index: 1.3770
Melting Point: -88°
Boiling Point: 78-80°
Beilstein Reference: 1737628
Merck Reference: 14,6112
UN No.: 1248
MDL No.: MFCD00009306
Formula:
C₄H₈O₂ Boiling Pt: 78…80 °C Melting Pt: –88 °C Density: 0.915 Flash Pt: –2 °C (28 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009306 CAS Number: 554-12-1 EINECS: 209-060-4 UN: 1248 ADR: 3,II Merck Index: 14,6112 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...