2-Acetylfuran 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14938-18
A14938-30
CAAAA14938-18EA
60.83
CAD
CAAAA14938-18
CAAAA14938-30
2-Acetylfuran 99%
2-Acetylfuran
Purity: 99%
CAS No.: 1192-62-7
Molecular Formula: C6H6O2
Formula Weight: 110.11
Storage Temperature: 2°C to 8°C
Flash Point: 71°(159°F)
Refractive Index: 1.5070
Melting Point: 27-30°
Boiling Point: 171-173°
Beilstein Reference: 107909
UN No.: 2811
MDL No.: MFCD00003242
Formula:
C₆H₆O₂ Boiling Pt: 171…173 °C Melting Pt: 27…30 °C Density: 1.098 Flash Pt: 71 °C(159 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003242 CAS Number: 1192-62-7 EINECS: 214-757-1 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...