Tetraoctylammonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14929-09 A14929-18 A14929-30
CAAAA14929-09EA 87.22 CAD
CAAAA14929-09 CAAAA14929-18 CAAAA14929-30
Tetraoctylammonium bromide 98+%
Tetraoctylammonium bromide

Purity: 98+% CAS No.: 14866-33-2 Molecular Formula: [CH3(CH2)7]4NBr Formula Weight: 546.81 Melting Point: 94-99° Beilstein Reference: 4169176

Formula: C₃₂H₆₈BrN
Melting Pt: 94…99 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011863
CAS Number: 14866-33-2
EINECS: 238-936-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR