Benzyl acetoacetate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14915-14
A14915-22
A14915-18
CAAAAA14915-14EA
50.56
CAD
CAAAAA14915-14
CAAAAA14915-22
CAAAA14915-18
CAAAA14915-30
Benzyl acetoacetate 97%
Benzyl acetoacetate
Purity: 97%
CAS No.: 5396-89-4
Molecular Formula: C11H12O3
Formula Weight: 192.22
Flash Point: 108°(226°F)
Refractive Index: 1.5120
Boiling Point: 276°
Beilstein Reference: 2049615
MDL No.: MFCD00008785
Formula:
C₁₁H₁₂O₃ Boiling Pt: 276 °C Density: 1,113 Flash Pt: 108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008785 CAS Number: 5396-89-4 EINECS: 226-416-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...