3-Phenylpropionic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Phenylpropanoic acid, Hydrocinnamic acid

A14908-22 A14908-36
CAAAA14908-22EA 112.02 CAD
CAAAA14908-22 CAAAA14908-36
3-Phenylpropionic acid 99%
3-Phenylpropionic acid

Purity: 99% CAS No.: 501-52-0 Molecular Formula: C6H5(CH2)2CO2H Formula Weight: 150.18 Flash Point: 110°(230°F) Melting Point: 47-50° Boiling Point: 280° Beilstein Reference: 907515 Merck Reference: 14,4784 MDL No.: MFCD00002771

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 280 °C
Melting Pt: 47…50 °C
Density: 1.071
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002771
CAS Number: 501-52-0
EINECS: 207-924-5
Merck Index: 14,4784

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR