Heptafluorobutyric acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid

A14907-36 A14907-14 A14907-22
CAAAAA14907-36EA 1189.74 CAD
CAAAAA14907-36 CAAAA14907-14 CAAAA14907-22
Heptafluorobutyric acid 99%
Heptafluorobutyric acid

Purity: 99% CAS No.: 375-22-4 Molecular Formula: C4HF7O2 Formula Weight: 214.04 Flash Point: None Refractive Index: 1.300 Odor: Unpleasant Melting Point: -18° Boiling Point: 120-121° Beilstein Reference: 1426882 UN No.: 3265 MDL No.: MFCD00004171

Formula: C₄HF₇O₂
MW: 214.04 g/mol
Boiling Pt: 120…121 °C
Melting Pt: –18 °C
Density: 1.648
Storage Temperature: Ambient
MDL Number: MFCD00004171
CAS Number: 375-22-4
EINECS: 206-786-3
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR