Adenine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

A14906-18 A14906-30 A14906-14 A14906-36
CAAAAA14906-18EA 237 CAD
CAAAAA14906-18 CAAAAA14906-30 CAAAA14906-14 CAAAA14906-36
Adenine 99%
Adenine

Purity: 99% CAS No.: 73-24-5 Molecular Formula: C5H5N5 Formula Weight: 135.13 Odor: Odorless Melting Point: 360° dec. Beilstein Reference: 5777 Merck Reference: 14,152 UN No.: 2811 MDL No.: MFCD00041790

Formula: C₅H₅N₅
MW: 135.13 g/mol
Melting Pt: >360 °C (dec.)
Density: 0.99741 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041790
CAS Number: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,152

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR