Formylhydrazine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Formic acid hydrazide, Formohydrazide, Formic hydrazide

A14901-14 A14901-22
CAAAA14901-14EA 95.43 CAD
CAAAA14901-14 CAAAA14901-22
Formylhydrazine 98%
Formylhydrazine

Purity: 98% CAS No.: 624-84-0 Molecular Formula: HCONHNH2 Formula Weight: 60.06 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 55-59° Beilstein Reference: 635759 MDL No.: MFCD00007589

Formula: CH₄N₂O
MW: 60.06 g/mol
Boiling Pt: 90 °C (0.7 -rr)
Melting Pt: 55…59 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007589
CAS Number: 624-84-0
EINECS: 210-867-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR