Isobutyl acetate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methylpropyl acetate

A14892-AK A14892-0F A14892-AP
CAAAAA14892-AKEA 42.61 CAD
CAAAAA14892-AK CAAAA14892-0F CAAAA14892-AP
Isobutyl acetate 98%
Isobutyl acetate

Purity: 98% CAS No.: 110-19-0 Molecular Formula: CH3CO2CH2CH(CH3)2 Formula Weight: 116.16 Flash Point: 18°(64°F) Physical Form: Liquid Density: 0.870 Refractive Index: 1.3895 Odor: Fruit-like Melting Point: -99° Boiling Point: 116-117° Beilstein Reference: 1741909 Merck Reference: 14,5130 UN No.: 1213 MDL No.: MFCD00008932

Formula: C₆H₁₂O₂
Boiling Pt: 116…117 °C
Melting Pt: –99 °C
Density: 0.870
Flash Pt: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008932
CAS Number: 110-19-0
EINECS: 203-745-1
UN: 1213
ADR: 3,II
Merck Index: 14,5130

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR