2-(4-Hydroxyphenyl)ethanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol

A14891-03 A14891-06 A14891-14
CAAAA14891-03EA 48.82 CAD
CAAAA14891-03 CAAAA14891-06 CAAAA14891-14
2-(4-Hydroxyphenyl)ethanol 98%
2-(4-Hydroxyphenyl)ethanol

Purity: 98% CAS No.: 501-94-0 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 89-92° Boiling Point: 139°/1mm MDL No.: MFCD00002902

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 139 °C (1 mmHg)
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00002902
CAS Number: 501-94-0
EINECS: 207-930-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR