3-Hydroxy-2-nitropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14887-09
A14887-18
A14887-30
CAAAA14887-09EA
68.41
CAD
CAAAA14887-09
CAAAA14887-18
CAAAA14887-30
3-Hydroxy-2-nitropyridine 98%
3-Hydroxy-2-nitropyridine
Purity: 98%
CAS No.: 15128-82-2
Molecular Formula: C5H4N2O3
Formula Weight: 140.10
Melting Point: 69-73°
Beilstein Reference: 124473
Formula:
C₅H₄N₂O₃ Melting Pt: 69…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006258 CAS Number: 15128-82-2 EINECS: 239-191-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...