4-Bromo-2,6-dichloroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14884-14
A14884-22
CAAAA14884-06EA
44.3
CAD
CAAAA14884-06
CAAAA14884-14
CAAAA14884-22
4-Bromo-2,6-dichloroaniline 98%
4-Bromo-2,6-dichloroaniline
Purity: 98%
CAS No.: 697-88-1
Molecular Formula: C6H4BrCl2N
Formula Weight: 240.92
Melting Point: 85-88°
Beilstein Reference: 2089687
Formula:
C₆H₄BrCl₂N MW: 240.91 g/mol Boiling Pt: 290 °C (1013 hPa) Melting Pt: 85…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025162 CAS Number: 697-88-1 EINECS: 211-809-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...