3-Amino-1H-1,2,4-triazole 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-1,2,4-triazole, Amitrole, 3-Amino-1H-1,2,4-triazole

A14871-0B A14871-18 A14871-30
CAAAA14871-0BEA 546.68 CAD
CAAAA14871-0B CAAAA14871-18 CAAAA14871-30
3-Amino-1H-1,2,4-triazole 96%
3-Amino-1H-1,2,4-triazole

Purity: 96% CAS No.: 61-82-5 Molecular Formula: C2H4N4 Formula Weight: 84.08 Physical Form: Powder Density: 1.138 Appearance: White to cream Odor: Odorless Melting Point: 147-153° Beilstein Reference: 107687 Merck Reference: 14,489 UN No.: 3077 MDL No.: MFCD00005230

Formula: C₂H₄N₄
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 147…153 °C
Density: 1.138
Storage Temperature: Ambient
MDL Number: MFCD00005230
CAS Number: 61-82-5
EINECS: 200-521-5
UN: 3077
ADR: "9",III
Merck Index: 14,489

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR