2,3,5-Trimethylpyrazine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14870-09
A14870-18
A14870-30
CAAAA14870-09EA
59.25
CAD
CAAAA14870-09
CAAAA14870-18
CAAAA14870-30
2,3,5-Trimethylpyrazine 99%
2,3,5-Trimethylpyrazine
Purity: 99%
CAS No.: 14667-55-1
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 54°(129°F)
Refractive Index: 1.5050
Boiling Point: 171-172°
Beilstein Reference: 2423
UN No.: 1993
MDL No.: MFCD00006145
Formula:
C₇H₁₀N₂ Boiling Pt: 171…172 °C Density: 0.979 Flash Pt: 54 °C (129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006145 CAS Number: 14667-55-1 EINECS: 238-712-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...