Tributylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

A14868-0D A14868-0F A14868-AP
CAAAA14868-0DEA 570.38 CAD
CAAAA14868-0D CAAAA14868-0F CAAAA14868-AP
Tributylamine 98%
Tributylamine

Purity: 98% CAS No.: 102-82-9 Molecular Formula: C12H27N Formula Weight: 185.36 Flash Point: 71°(159°F) Refractive Index: 1.4290 Odor: Amine-like pH: 10.6 Melting Point: -70° Boiling Point: 216° Beilstein Reference: 1698872 Merck Reference: 14,9618 UN No.: 2542 MDL No.: MFCD00009431

Formula: C₁₂H₂₇N
Boiling Pt: 216 °C
Melting Pt: < –70 °C
Density: 0.778
Flash Pt: 71 °C(159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009431
CAS Number: 102-82-9
EINECS: 203-058-7
UN: 2542
ADR: 6.1,II
Merck Index: 14,9618

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR