2-Bromo-4-nitrotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14865-14
A14865-22
CAAAA14865-14EA
91.96
CAD
CAAAA14865-14
CAAAA14865-22
2-Bromo-4-nitrotoluene 98%
2-Bromo-4-nitrotoluene
Purity: 98%
CAS No.: 7745-93-9
Molecular Formula: C7H6BrNO2
Formula Weight: 216.04
Melting Point: 74-78°
Beilstein Reference: 1941232
Formula:
C₇H₆BrNO₂ Boiling Pt: 175 °C (38 -rr) Melting Pt: 74…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007195 CAS Number: 7745-93-9 EINECS: 231-809-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...