1,4-Diphenoxybenzene 98%

Supplier: Thermo Scientific Chemicals

A14864-22 A14864-06 A14864-14
CAAAAA14864-22EA 944.84 CAD
CAAAAA14864-22 CAAAA14864-06 CAAAA14864-14
1,4-Diphenoxybenzene 98%
1,4-Diphenoxybenzene

Purity: 98% CAS No.: 3061-36-7 Molecular Formula: C18H14O2 Formula Weight: 262.31 Flash Point: 202°(395°F) Melting Point: 72-73° Boiling Point: 172°/0.01mm Beilstein Reference: 2215945 MDL No.: MFCD00038368

Formula: C₁₈H₁₄O₂
Boiling Pt: 172 °C/0.01 mm
Melting Pt: 72…73 °C
Density: 1.083
Flash Pt: 202 °C (395 °F)
Storage Temperature: Ambient
MDL Number: MFCD00038368
CAS Number: 3061-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR