Ethyl valerate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl Pentanoate, Valeric acid ethyl ester

A14854-0F A14854-AP
CAAAA14854-0FEA 445.56 CAD
CAAAA14854-0F CAAAA14854-AP
Ethyl valerate 98%
Ethyl valerate

Purity: 98% CAS No.: 539-82-2 Molecular Formula: CH3(CH2)3CO2CH2CH3 Formula Weight: 130.19 Flash Point: 38°(100°F) Refractive Index: 1.400 Melting Point: -92 to -90° Boiling Point: 142-145° Beilstein Reference: 1744680 Merck Reference: 14,9904 UN No.: 3272 MDL No.: MFCD00009479

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 145
Melting Pt: -91
Density: 0.874
Flash Pt: 38 °C (100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009479
CAS Number: 539-82-2
EINECS: 208-726-1
UN: 3272
ADR: 3 III
Merck Index: 14,9904

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR