Pyrrolidine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00005249
Beilstein: 102395
Soluble in alcohol, ether, chloroform. Miscible with water
Purity: 99%
CAS No.: 123-75-1
Molecular Formula: C4H9N
Formula Weight: 71.12
Flash Point: 2°(35°F)
Physical Form: Liquid
Density: 0.856
Refractive Index: 1.4431
Sensitivity: Air Sensitive
Appearance: Colorless to yellow
Odor: Like rotten eggs
Melting Point: -63°
Boiling Point: 86-88°
Solubility: Soluble in alcohol, ether, chloroform. Miscible with water
Beilstein Reference: 102395
Merck Reference: 14,8015
UN No.: 1922
MDL No.: MFCD00005249
Formula:
C₄H₉N Boiling Pt: 86…88 °C Melting Pt: –63 °C Density: 0.856 Flash Pt: 2 °C (35 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005249 CAS Number: 123-75-1 EINECS: 204-648-7 UN: 1922 ADR: 3,II Merck Index: 14,8015 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...