Nicotinonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Pyridinecarbonitrile, 3-Cyanopyridine, Pyridine-3-carbonitrile
A14850-0E
A14850-22
A14850-36
CAAAA14850-0EEA
680.98
CAD
CAAAA14850-0E
CAAAA14850-22
CAAAA14850-36
Nicotinonitrile 98%
Nicotinonitrile
Purity: 98%
CAS No.: 100-54-9
Molecular Formula: C6H4N2
Formula Weight: 104.11
Flash Point: 84°(183°F)
Odor: Pungent
Boiling Point: 206-208°
Beilstein Reference: 107711
MDL No.: MFCD00006372
Formula:
C₆H₄N₂ Boiling Pt: 206…208 °C Melting Pt: 48…51 °C Density: 1.16 Flash Pt: 84 °C(183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006372 CAS Number: 100-54-9 EINECS: 202-863-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...