2-Amino-4-methylthiazole 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methylthiazol-2-ylamine, 4-Methylthiazol-2-amine, 2-Amino-4-methylthiazol

A14847-06 A14847-14 A14847-22
CAAAA14847-06EA 34.6 CAD
CAAAA14847-06 CAAAA14847-14 CAAAA14847-22
2-Amino-4-methylthiazole 98+%
2-Amino-4-methylthiazole

Purity: 98+% CAS No.: 1603-91-4 Molecular Formula: C4H6N2S Formula Weight: 114.17 Flash Point: 107°(224°F) Odor: Characteristic Boiling Point: 231-233° Beilstein Reference: 107890 Merck Reference: 14,450 MDL No.: MFCD00005329

Formula: C₄H₆N₂S
MW: 114.17 g/mol
Boiling Pt: 231…233 °C
Melting Pt: 42…46 °C
Density: 1.202 g/cm³ (50 °C)
Flash Pt: 107 °C(224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005329
CAS Number: 1603-91-4
EINECS: 216-505-6
Merck Index: 14,450

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR