Thioanisole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl phenyl sulphide, Methyl phenyl sulfide, Phenyl methyl sulphide, Phenyl methyl sulfide, (Methylthio)benzene

A14846-14 A14846-0E A14846-22 A14846-36
CAAAAA14846-14EA 53.25 CAD
CAAAAA14846-14 CAAAA14846-0E CAAAA14846-22 CAAAA14846-36
Thioanisole 99%
Thioanisole

Purity: 99% CAS No.: 100-68-5 Molecular Formula: C7H8S Formula Weight: 124.21 Flash Point: 57°(135°F) Refractive Index: 1.5860 Odor: Stench Melting Point: -15° Boiling Point: 188-189° Beilstein Reference: 1904179 UN No.: 1993 MDL No.: MFCD00008559

Formula: C₇H₈S
MW: 124.21 g/mol
Boiling Pt: 188
Melting Pt: -15
Density: 1.058
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008559
CAS Number: 100-68-5
EINECS: 202-878-2
UN: 3082
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR