2,6-Dichlorobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,6-Dichlorobenzoyl Chloride

A14839-09 A14839-18
CAAAA14839-09EA 80.58 CAD
CAAAA14839-09 CAAAA14839-18
2,6-Dichlorobenzoyl chloride 98%
2,6-Dichlorobenzoyl chloride

Purity: 98% CAS No.: 4659-45-4 Molecular Formula: C7H3Cl3O Formula Weight: 209.46 Flash Point: 110°(230°F) Refractive Index: 1.5600 Melting Point: 15-18° Boiling Point: 130-132°/15mm Beilstein Reference: 639531 UN No.: 3265 MDL No.: MFCD00000662

Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 130…132 °C/15 mm
Melting Pt: 15…18 °C
Density: 1.463
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000662
CAS Number: 4659-45-4
EINECS: 225-102-4
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR