2-Bromothiazole 99%

Supplier: Thermo Scientific Chemicals
Warning

A14838-14 A14838-09 A14838-18
CAAAAA14838-14EA 136.51 CAD
CAAAAA14838-14 CAAAA14838-09 CAAAA14838-18 CAAAA14838-30
2-Bromothiazole 99%
2-Bromothiazole

Purity: 99% CAS No.: 3034-53-5 Molecular Formula: C3H2BrNS Formula Weight: 164.02 Storage Temperature: 2°C to 8°C Flash Point: 63°(145°F) Refractive Index: 1.5930 Boiling Point: 171° Beilstein Reference: 105724 MDL No.: MFCD00005316

Formula: C₃H₂BrNS
MW: 164.03 g/mol
Boiling Pt: 171 °C
Density: 1,836
Flash Pt: 63 °C(145 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005316
CAS Number: 3034-53-5
EINECS: 221-229-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR