n-Dodecane 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14834-0F
A14834-AE
A14834-AP
CAAAA14834-0FEA
818.44
CAD
CAAAA14834-0F
CAAAA14834-AE
CAAAA14834-AP
n-Dodecane 99+%
n-Dodecane
Purity: 99+%
CAS No.: 112-40-3
Molecular Formula: CH3(CH2)10CH3
Formula Weight: 170.34
Flash Point: 71°(159°F)
Physical Form: Liquid
Density: 0.749
Refractive Index: 1.4220
Odor: Petrol-like
Melting Point: -10°
Boiling Point: 215-217°
Beilstein Reference: 1697175
MDL No.: MFCD00008969
Formula:
H₃C(CH₂)₁₀CH₃ Boiling Pt: 215…217 °C Melting Pt: –10 °C Density: 0.749 Flash Pt: 71 °C(159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008969 CAS Number: 112-40-3 EINECS: 203-967-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...