(4-Nitrophenyl)acetonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

A14821-18 A14821-30
CAAAA14821-18EA 82 CAD
CAAAA14821-18 CAAAA14821-30
(4-Nitrophenyl)acetonitrile 98%
(4-Nitrophenyl)acetonitrile

Purity: 98% CAS No.: 555-21-5 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: 114-117° Boiling Point: 195-197°/12mm Merck Reference: 14,6590 UN No.: 3439 MDL No.: MFCD00007372

Formula: C₈H₆N₂O₂
Boiling Pt: 195…197 °C/12 mm
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00007372
CAS Number: 555-21-5
EINECS: 209-085-0
UN: 3439
ADR: 6.1,III
Merck Index: 14,6590

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR